Learn More
Research Products International Corp Thiamphenicol, 5 G
SDP
Catalog No. 501977864
Encompass_Preferred
Thiamphenicol is a compound which plays a pivotal role in the study of bacterial infections and the development of antibiotic therapies.
In microbiology, Thiamphenicol is a valuable tool for investigating bacterial susceptibility and resistance patterns. It serves as a reference standard for assessing the effectiveness of antibiotics against various bacterial strains, shedding light on the mechanisms underlying antibiotic resistance. This research deepens our understanding of microbial biology and informs strategies for combating bacterial infections. 
Pharmaceutical researchers rely on Thiamphenicol to develop and optimize antibacterial therapies. Its use in testing and formulation contributes to the fight against antibiotic-resistant bacteria, a growing concern in the field of medicine. 
Catalog No. 50-197-7864 Supplier Research Products International Corp Supplier No. T561505.0
Thiamphenicol is a compound which plays a pivotal role in the study of bacterial infections and the development of antibiotic therapies.
In microbiology, Thiamphenicol is a valuable tool for investigating bacterial susceptibility and resistance patterns. It serves as a reference standard for assessing the effectiveness of antibiotics against various bacterial strains, shedding light on the mechanisms underlying antibiotic resistance. This research deepens our understanding of microbial biology and informs strategies for combating bacterial infections. 
Pharmaceutical researchers rely on Thiamphenicol to develop and optimize antibacterial therapies. Its use in testing and formulation contributes to the fight against antibiotic-resistant bacteria, a growing concern in the field of medicine. 