Learn More
Apexbio Technology LLC Thiamphenicol 15318-45-3 50mg
SDP
Catalog No. 502240636
Encompass_Preferred
Thiamphenicol (CAS 15318-45-3) is a small-molecule inhibitor targeting the bacterial ribosomal 50S subunit It is designed to interfere with bacterial protein synthesis thereby regulating translational processes Thiamphenicol exerts its biological activity primarily through inhibition of peptide elongation by binding to the 50S ribosomal subunit In experimental studies Thiamphenicol demonstrates bacteriostatic activity with reported IC50 values for bacterial strains typically ranging from 0 5 8 g/mL depending on organism sensitivity growth conditions and assay methodologies Based on these pharmacological properties Thiamphenicol holds research potential in studying bacterial resistance mechanisms screening antibiotic candidates and investigating translation regulation in microbial systems
This item is not returnable. View return policy
Catalog No. 50-224-0636 Supplier Apexbio Technology LLC Supplier No. B185350
Thiamphenicol (CAS 15318-45-3) is a small-molecule inhibitor targeting the bacterial ribosomal 50S subunit It is designed to interfere with bacterial protein synthesis thereby regulating translational processes Thiamphenicol exerts its biological activity primarily through inhibition of peptide elongation by binding to the 50S ribosomal subunit In experimental studies Thiamphenicol demonstrates bacteriostatic activity with reported IC50 values for bacterial strains typically ranging from 0 5 8 g/mL depending on organism sensitivity growth conditions and assay methodologies Based on these pharmacological properties Thiamphenicol holds research potential in studying bacterial resistance mechanisms screening antibiotic candidates and investigating translation regulation in microbial systems