Learn More
Cayman Chemical ThIamphenIcol 5g
SDP
Catalog No. 502981490
Encompass_Preferred
An antibiotic derived from chloramphenicol that displays a similar spectrum of activity with enhanced potency
Catalog No. 50-298-1490 Supplier Cayman Chemical Supplier No. 213575 G
An antibiotic derived from chloramphenicol that displays a similar spectrum of activity with enhanced potency