Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamphenicol, 99.5%, MP Biomedicals™

Catalog No. ICN15684605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15684605 5 g
ICN15684625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15684605 Supplier MP Biomedicals, Inc Supplier No. 0215684605
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15318-45-3
Molecular Formula C12H15Cl2NO5S
Molecular Weight (g/mol) 356.214
InChI Key OTVAEFIXJLOWRX-NXEZZACHSA-N
Synonym D -threo-2, 2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-(methyl- sulfonyl)phenethyl]-acetamide
PubChem CID 27200
ChEBI CHEBI:32215
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O

Specifications

Type Thiamphenicol
Physical Form Free-flowing Powder
Color White
Quantity 5 g
Assay Percent Range 0.995
Formula Weight 356.2
Chemical Name or Material Thiamphenicol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.