Learn More
Medchemexpress LLC Thiamphenicol-d3-1 | 1217723-41-5 | 99.0% | 1 MG
SDP
Catalog No. 5000407812
Encompass_Preferred
Thiamphenicol-d3-1 is a deuterium-labeled form of Thiamphenicol, a methyl-sulfonyl derivative of Chloramphenicol. It acts as a broad-spectrum antimicrobial antibiotic by binding to the 50S ribosomal subunit, inhibiting protein synthesis and resulting in a bacteriostatic effect against Gram-negative, Gram-positive aerobic, and anaerobic bacteria. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs, and stable heavy isotopes are incorporated for quantitation during drug development.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits broad-spectrum antimicrobial activity
  • Inhibits protein synthesis by binding to the 50S ribosomal subunit
  • Effective against Gram-negative, Gram-positive aerobic, and anaerobic bacteria
Catalog No. 50-004-07812 Supplier Medchemexpress LLC Supplier No. HYB0479S11MG
Thiamphenicol-d3-1 is a deuterium-labeled form of Thiamphenicol, a methyl-sulfonyl derivative of Chloramphenicol. It acts as a broad-spectrum antimicrobial antibiotic by binding to the 50S ribosomal subunit, inhibiting protein synthesis and resulting in a bacteriostatic effect against Gram-negative, Gram-positive aerobic, and anaerobic bacteria. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs, and stable heavy isotopes are incorporated for quantitation during drug development.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits broad-spectrum antimicrobial activity
  • Inhibits protein synthesis by binding to the 50S ribosomal subunit
  • Effective against Gram-negative, Gram-positive aerobic, and anaerobic bacteria