Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiamphenicol, G-Biosciences
SCI_ED

Catalog No. S139830
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139830 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139830 Supplier G-Biosciences Supplier No. RC1115
Add to Cart
Add to Cart

Thiamphenicol is a bacteriostatic antibiotic derived from chloramphenicol. It is effective against several gram-positive and gram-negative bacteria

Thiamphenicol commonly used in researching protein synthesis and to select for chloramphenicol-resistant transformed cells or the bacterial CAT gene. It can be used against uncomplicated gonorrhea infections and chancroid.

Chemical Identifiers

CAS 15318-45-3

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 5 g
Percent Purity 98%
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.