Learn More
Medchemexpress LLC Thiamphenicol (Standard) | 15318-45-3 | 99.6% | 50 MG
SDP
Catalog No. 5000409427
Encompass_Preferred
Thiamphenicol (Standard) is an analytical standard and a methyl-sulfonyl derivative of Chloramphenicol. This broad-spectrum antimicrobial antibiotic inhibits protein synthesis by binding to the 50S ribosomal subunit, providing a bacteriostatic effect against Gram-negative, Gram-positive aerobic, and anaerobic bacteria. It is intended for research and analytical applications.
  • Analytical standard
  • Methyl-sulfonyl derivative of chloramphenicol
  • Broad-spectrum antimicrobial activity
  • Inhibits bacterial protein synthesis
  • Effective against Gram-negative, Gram-positive aerobic, and anaerobic bacteria
  • Suitable for research and analytical applications
Catalog No. 50-004-09427 Supplier Medchemexpress LLC Supplier No. HYB0479R50MG
Thiamphenicol (Standard) is an analytical standard and a methyl-sulfonyl derivative of Chloramphenicol. This broad-spectrum antimicrobial antibiotic inhibits protein synthesis by binding to the 50S ribosomal subunit, providing a bacteriostatic effect against Gram-negative, Gram-positive aerobic, and anaerobic bacteria. It is intended for research and analytical applications.
  • Analytical standard
  • Methyl-sulfonyl derivative of chloramphenicol
  • Broad-spectrum antimicrobial activity
  • Inhibits bacterial protein synthesis
  • Effective against Gram-negative, Gram-positive aerobic, and anaerobic bacteria
  • Suitable for research and analytical applications