Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiamphenicol (standard) | 15318-45-3 | 99.8% | 25 MG
SDP

Catalog No. 5000451025
Encompass_Preferred

Thiamphenicol (Standard) is the analytical standard of Thiamphenicol, a broad-spectrum antimicrobial antibiotic. It is a methyl-sulfonyl derivative of Chloramphenicol. This product is intended for research and analytical applications.

  • Mechanism of action: Thiamphenicol binds to the 50S ribosomal subunit, which inhibits protein synthesis and provides a bacteriostatic effect against gram-negative, gram-positive aerobic, and anaerobic bacteria.
  • Application: Used as a reference standard assay in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Appearance: Solid, white to off-white.
  • Molecular weight: 356.22.
  • For research use only.

Catalog No. 50-004-51025 Supplier Medchemexpress LLC Supplier No. 25MGHYB0479R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiamphenicol (Standard) is the analytical standard of Thiamphenicol, a broad-spectrum antimicrobial antibiotic. It is a methyl-sulfonyl derivative of Chloramphenicol. This product is intended for research and analytical applications.

  • Mechanism of action: Thiamphenicol binds to the 50S ribosomal subunit, which inhibits protein synthesis and provides a bacteriostatic effect against gram-negative, gram-positive aerobic, and anaerobic bacteria.
  • Application: Used as a reference standard assay in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Appearance: Solid, white to off-white.
  • Molecular weight: 356.22.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.