Learn More
Medchemexpress LLC Thianthrene | 92-85-3 | MFCD00005065 | 100.0% | 216.32 g/mol | C12H8S2 | 25 G
SDP
Catalog No. 5000303511
Encompass_Preferred
Thianthrene (CAS 92-85-3) is a sulfur-containing tricyclic research chemical used in biochemical assays and reported as an inhibitor of Leishmania donovani pteridine reductase 1 (PTR1). It is supplied for research and analytical applications with high reported purity and is available in solution and solid forms to support various laboratory workflows.
  • High purity suitable for biochemical research.
  • Reported PTR1 inhibitory activity for parasite biology studies.
  • Available as DMSO solution and solid quantities for experimental flexibility.
  • Intended for research and analytical applications only.
  • Provided with a safety data sheet and handling documentation.
Catalog No. 50-003-03511 Supplier Medchemexpress LLC Supplier No. 25GHYW014120
Thianthrene (CAS 92-85-3) is a sulfur-containing tricyclic research chemical used in biochemical assays and reported as an inhibitor of Leishmania donovani pteridine reductase 1 (PTR1). It is supplied for research and analytical applications with high reported purity and is available in solution and solid forms to support various laboratory workflows.
  • High purity suitable for biochemical research.
  • Reported PTR1 inhibitory activity for parasite biology studies.
  • Available as DMSO solution and solid quantities for experimental flexibility.
  • Intended for research and analytical applications only.
  • Provided with a safety data sheet and handling documentation.