Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Thianthrene S-Oxide

Catalog No. 9039735G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-397-35G 1 g
90-397-31G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-397-35G Supplier Sigma Aldrich Supplier No. 9039735G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Thianthrene S-oxide is a sulfoxide-based thianthrene reagent for late-stage C-H functionalization. Developed by the Ritter Lab, C-H functionalization by thianthrenation proceeds with >99% selectivity and furnishes functionalized arenes serving as synthetic linchpins for further participation in diverse transformations. Nonfluorinated thianthrene S-oxide is best used for all arenes more electron-rich than anisole. For other arenes, the fluorinated TFT S-oxide (903981) was demonstrated as the thanthrenation reagent.Site-selective and versatile aromatic C-H functionalization by thianthrenationVideo: Site-selective C-H functionalization by thianthrenationIn the Pipeline: New Chemistry, Making New ThingsTi(Phen)(OC2H5)2Cl2: a highly efficient pre-?catalyst for selective oxidation of organic sulfides to sulfoxides by hydrogen peroxidePatent Application EP18204755.5 This product is manufactured pursuant to a license with Studiengesellschaft Kohle mbH

Specifications

CAS 23570-43-6
Molecular Formula C12H8OS2
Linear Formula C12H8OS2
MDL Number MFCD01464038
Quantity 1 g
Synonym Non-fluorinated sulfoxide-based thianthrene reagent; Thianthrene 5-oxide
Molecular Weight (g/mol) 232.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.