Learn More
AdipoGen Thiazole Orange, 107091-89-4, MFCD00192499, 5 g
SDP
Catalog No. 50595594
Encompass_Preferred
MF: C26H24N2O3S2, MW: 476.61, ≥98%, Appearance: Orange crystalline powder. Thiazol Orange is a nucleic acid stain that has found applications in many different areas. Among others it is used for reticulocyte analysis and Plasmodium species analysis as well as labeling cancer cells. Thiazole orange-labeled DNA has been used for live cell RNA imaging. Thiazole Orange has also been used for a variety of assays, PCR and flow cytometry.
Catalog No. 50-595-594 Supplier AdipoGen Supplier No. CDXT0013G005
MF: C26H24N2O3S2, MW: 476.61, ≥98%, Appearance: Orange crystalline powder. Thiazol Orange is a nucleic acid stain that has found applications in many different areas. Among others it is used for reticulocyte analysis and Plasmodium species analysis as well as labeling cancer cells. Thiazole orange-labeled DNA has been used for live cell RNA imaging. Thiazole Orange has also been used for a variety of assays, PCR and flow cytometry.