Learn More
Abcam Thiazol Orange, Fluorescent dye for reticulocyte analysis, 250MG
Catalog No. 502705331
Encompass_Preferred
MW 476.6 Da, Purity >90%. Fluorescent dye for reticulocyte analysis. Nucleic acid stain with high affinity for dsDNA (log K = 5.5 at 100 mM salt). Cell-permeable without pretreatment. Suitable for live cell RNA imaging.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5331 Supplier Abcam Supplier No. AB145287250MG
MW 476.6 Da, Purity >90%. Fluorescent dye for reticulocyte analysis. Nucleic acid stain with high affinity for dsDNA (log K = 5.5 at 100 mM salt). Cell-permeable without pretreatment. Suitable for live cell RNA imaging.
The product is subject to the following: Abcam Restricted Use Statement