Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiazole Orange | 107091-89-4 | 99.6% | 10 MM 1 ML
SDP

Catalog No. 5000410814
Encompass_Preferred

Thiazole Orange | 107091-89-4 | 99.6% | 10 MM 1 ML

Catalog No. 50-004-10814 Supplier Medchemexpress LLC Supplier No. HYD015010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiazole Orange | 107091-89-4 | 99.6% | 10 MM 1 ML

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.