Learn More
Medchemexpress LLC Thiazole orange dimer YOYO 1 | 143413-85-8 | 97.0% | 635 UG
SDP
Catalog No. 5000410205
Encompass_Preferred
Thiazole orange dimer YOYO 1 is a green fluorescent dye utilized for DNA staining. It is a monomethine cyanine dye, presenting as a tetracationic homodimer of oxazole yellow (YO), and is typically supplied as a tetraiodide salt. While exhibiting low fluorescence in aqueous buffer, its intensity significantly increases upon binding to double-stranded DNA through diintercalation.
  • Green fluorescent dye
  • Used for DNA staining
  • Monomethine cyanine dye family
  • Tetracationic homodimer of oxazole yellow
  • Fluorescence increases 3200 times upon binding to double-stranded DNA
Catalog No. 50-004-10205 Supplier Medchemexpress LLC Supplier No. HYD0916635UG
Thiazole orange dimer YOYO 1 is a green fluorescent dye utilized for DNA staining. It is a monomethine cyanine dye, presenting as a tetracationic homodimer of oxazole yellow (YO), and is typically supplied as a tetraiodide salt. While exhibiting low fluorescence in aqueous buffer, its intensity significantly increases upon binding to double-stranded DNA through diintercalation.
  • Green fluorescent dye
  • Used for DNA staining
  • Monomethine cyanine dye family
  • Tetracationic homodimer of oxazole yellow
  • Fluorescence increases 3200 times upon binding to double-stranded DNA