Learn More
AdipoGen Thiazole Orange Solution
SDP
Catalog No. 502559936
Encompass_Preferred
Chemical. CAS 107091-89-4. Formula C26H24N2O3S2. MW 476.61. Thiazol Orange is a nucleic acid stain that has found applications in many different areas. Among others it is used for reticulocyte analysis and Plasmodium species analysis as well as labeling cancer cells. Thiazole orange-labeled DNA has been used for live cell RNA imaging. Thiazole Orange has also been used for a variety of assays, PCR and flow cytometry. Spectral data lambdaex=509nm, lambdaem=530nm in 0.1M phosphate pH 7.0 bound to dsDNA.
Catalog No. 50-255-9936 Supplier AdipoGen Supplier No. CDXT0513M025
Chemical. CAS 107091-89-4. Formula C26H24N2O3S2. MW 476.61. Thiazol Orange is a nucleic acid stain that has found applications in many different areas. Among others it is used for reticulocyte analysis and Plasmodium species analysis as well as labeling cancer cells. Thiazole orange-labeled DNA has been used for live cell RNA imaging. Thiazole Orange has also been used for a variety of assays, PCR and flow cytometry. Spectral data lambdaex=509nm, lambdaem=530nm in 0.1M phosphate pH 7.0 bound to dsDNA.