Learn More
Biotium Thiazole Red, 1mM in DMSO
Catalog No. 501964665
Encompass_Preferred
Thiazole red, also known as TO-PRO-3, is a cell-impermeant, high-affinity carbocyanine monomeric nucleic acid stain that is essentially nonfluorescent in the absence of nucleic acids but exhibits excitation/emission maxima ~642/661 nm when bound to nucleic acids. Thiazole red is also used as a nuclear counterstain and dead cell indicator. Properties: Blue solution; Ex/Em= 642/661 nm (with DNA); Store at -20°C and protect from light; C26H31I2N3S, MW: 671.42.
Catalog No. 50-196-4665 Supplier Biotium Supplier No. 40087
 Restricted Item
Add to Cart
Add to Cart
Thiazole red, also known as TO-PRO-3, is a cell-impermeant, high-affinity carbocyanine monomeric nucleic acid stain that is essentially nonfluorescent in the absence of nucleic acids but exhibits excitation/emission maxima ~642/661 nm when bound to nucleic acids. Thiazole red is also used as a nuclear counterstain and dead cell indicator. Properties: Blue solution; Ex/Em= 642/661 nm (with DNA); Store at -20°C and protect from light; C26H31I2N3S, MW: 671.42.