Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thiazole 98.0+%, TCI America™
SDP

Catalog No. T018525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T018525G 25 g
T01855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T018525G Supplier TCI America Supplier No. T018525G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 288-47-1
Molecular Formula C3H3NS
Molecular Weight (g/mol) 85.12
MDL Number MFCD00005315
InChI Key FZWLAAWBMGSTSO-UHFFFAOYSA-N
Synonym thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium
PubChem CID 9256
ChEBI CHEBI:43732
IUPAC Name 1,3-thiazole
SMILES S1C=CN=C1

Specifications

Melting Point -33°C
Color Yellow
Boiling Point 118°C
Quantity 25 g
UN Number 1993
Formula Weight 85.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Thiazole
RTECSNumber XJ1290000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.