Learn More
Medchemexpress LLC Thiazole Yellow G f 100g
SDP
Catalog No. 5000206333
Encompass_Preferred
Thiazole yellow G for microscopy is a cationic basic thiazine dye Thiazole yellow G can be used for the determination of uranium and thorium[1][2]
Catalog No. 50-002-06333 Supplier Medchemexpress LLC Supplier No. HYW110909100G
Thiazole yellow G for microscopy is a cationic basic thiazine dye Thiazole yellow G can be used for the determination of uranium and thorium[1][2]