Learn More
Medchemexpress LLC Thiazole Yellow G f 50g
SDP
Catalog No. 5000206345
Encompass_Preferred
Thiazole yellow G for microscopy is a cationic basic thiazine dye Thiazole yellow G can be used for the determination of uranium and thorium[1][2]
Catalog No. 50-002-06345 Supplier Medchemexpress LLC Supplier No. HYW11090950G
Thiazole yellow G for microscopy is a cationic basic thiazine dye Thiazole yellow G can be used for the determination of uranium and thorium[1][2]