Learn More
Medchemexpress LLC Thiazolidine | 504-78-9 | 99.78% | 89.16 | 500 G
SDP
Catalog No. 5000431824
Encompass_Preferred
Thiazolidine is a five-membered heterocyclic compound that contains both sulfur and nitrogen heteroatoms. It serves as a crucial intermediate in organic synthesis. Derivatives of Thiazolidine have demonstrated a wide array of biological activities, including anti-tumor, antibacterial, and insecticidal properties, as well as the ability to lower blood glucose levels.
  • Five-membered heterocyclic compound with sulfur and nitrogen heteroatoms
  • Crucial intermediate in organic synthesis
  • Derivatives show anti-tumor activity
  • Derivatives show antibacterial activity
  • Derivatives show insecticidal properties
  • Derivatives show ability to lower blood glucose levels
  • Intended for research use only
Catalog No. 50-004-31824 Supplier Medchemexpress LLC Supplier No. HYI0144500G
Thiazolidine is a five-membered heterocyclic compound that contains both sulfur and nitrogen heteroatoms. It serves as a crucial intermediate in organic synthesis. Derivatives of Thiazolidine have demonstrated a wide array of biological activities, including anti-tumor, antibacterial, and insecticidal properties, as well as the ability to lower blood glucose levels.
  • Five-membered heterocyclic compound with sulfur and nitrogen heteroatoms
  • Crucial intermediate in organic synthesis
  • Derivatives show anti-tumor activity
  • Derivatives show antibacterial activity
  • Derivatives show insecticidal properties
  • Derivatives show ability to lower blood glucose levels
  • Intended for research use only