Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiazolidine hydrochloride | 14446-47-0 | 95.0% | C3H8ClNS | 100 G
SDP

Catalog No. 5000435002
Encompass_Preferred

Thiazolidine hydrochloride is a chemical substance primarily used as a drug intermediate.

  • Appears as a white to yellow solid
  • Molecular formula is C3H8ClNS
  • Molecular weight is 125.62
  • Purity (NMR) is 95.0%

Catalog No. 50-004-35002 Supplier Medchemexpress LLC Supplier No. HY77599100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiazolidine hydrochloride is a chemical substance primarily used as a drug intermediate.

  • Appears as a white to yellow solid
  • Molecular formula is C3H8ClNS
  • Molecular weight is 125.62
  • Purity (NMR) is 95.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.