Learn More
Medchemexpress LLC Thiazolidine hydrochloride | 14446-47-0 | 95.0% | C3H8ClNS | 25 G
SDP
Catalog No. 5000359176
Encompass_Preferred
Thiazolidine hydrochloride is a chemical compound identified by CAS number 14446-47-0 and molecular formula C3H8ClNS. It appears as a white to yellow solid with an NMR purity of 95.0% and is stable under recommended storage conditions. This compound is suitable for various laboratory applications and the manufacture of other substances.
  • Used as a laboratory chemical.
  • Suitable for manufacturing various substances.
  • Stable under recommended storage conditions.
  • White to yellow solid appearance.
  • Molecular formula C3H8ClNS.
Catalog No. 50-003-59176 Supplier Medchemexpress LLC Supplier No. 25GHY77599
Thiazolidine hydrochloride is a chemical compound identified by CAS number 14446-47-0 and molecular formula C3H8ClNS. It appears as a white to yellow solid with an NMR purity of 95.0% and is stable under recommended storage conditions. This compound is suitable for various laboratory applications and the manufacture of other substances.
  • Used as a laboratory chemical.
  • Suitable for manufacturing various substances.
  • Stable under recommended storage conditions.
  • White to yellow solid appearance.
  • Molecular formula C3H8ClNS.