Learn More
Selleck Chemical LLC Thiazolidinedione-E1775-100G
SDP
Catalog No. 503138891
Encompass_Preferred
2 4-Thiazolidinedione (Thiazolidinedione) an insulin sensitizer is a specific agonist of peroxisome proliferator-activated receptor (PPAR) Thiazolidinedione treatment corrects impaired muscle insulin action in vivo
Catalog No. 50-313-8891 Supplier Selleck Chemical LLC Supplier No. E1775100G
2 4-Thiazolidinedione (Thiazolidinedione) an insulin sensitizer is a specific agonist of peroxisome proliferator-activated receptor (PPAR) Thiazolidinedione treatment corrects impaired muscle insulin action in vivo