Learn More
Medchemexpress LLC Thiazolo[4,5-d]pyrimidine-2(3H)-thione, 7-chloro-3-(2,4-dimethoxyphenyl)-5-methyl- | 2459916-56-2 | 98.5% | 353.85 | 25 MG
SDP
Catalog No. 5000363267
Encompass_Preferred
CDK1/Cyc B-IN-1 (Compound 5) is a selective CDK1/Cyc B complex inhibitor with an IC50 of 97 nM. It induces apoptosis and G2/M cell cycle arrest, and exhibits broad-spectrum cytotoxic action against various cancer cell lines.
  • Selective inhibitor of the CDK1/Cyc B complex.
  • Induces apoptosis and G2/M cell cycle arrest.
  • Exhibits broad-spectrum cytotoxic action against cancer cell lines.
  • Cytotoxic activity against HCT-116 cells and WI-38 cells.
Catalog No. 50-003-63267 Supplier Medchemexpress LLC Supplier No. 25MGHY147646
CDK1/Cyc B-IN-1 (Compound 5) is a selective CDK1/Cyc B complex inhibitor with an IC50 of 97 nM. It induces apoptosis and G2/M cell cycle arrest, and exhibits broad-spectrum cytotoxic action against various cancer cell lines.
  • Selective inhibitor of the CDK1/Cyc B complex.
  • Induces apoptosis and G2/M cell cycle arrest.
  • Exhibits broad-spectrum cytotoxic action against cancer cell lines.
  • Cytotoxic activity against HCT-116 cells and WI-38 cells.