Learn More
Medchemexpress LLC Thiazolo[5,4-e]triazolo[1,5-c]pyrimidin-2(3h)-one, 5-amino-3-[2-[4-[2,4-difluoro-5-[2-[(s)-methylsulfinyl]ethoxy]phenyl]-1-piperazinyl]ethyl]-8-(2-furanyl)-, hydrochloride (1:1) | 2411004-22-1 | 99.6% | 641.11 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY137442A
Encompass_Preferred
Inupadenant hydrochloride (EOS-850) is an orally active, highly selective A2A receptor antagonist that cannot cross the blood-brain barrier. It enhances the humoral immune response and exhibits anti-tumor activity.
Catalog No. 50-003-47788