Learn More
Medchemexpress LLC Thiazolo[5,4-f]quinazoline-2-carboximidic acid, 9-[(2-fluoro-4-methoxyphenyl)amino]-, methyl ester | 1425945-60-3 | 98.9% | 383.40 | 50 MG
SDP
Catalog No. 5000343327
Encompass_Preferred
EHT 1610 is a potent inhibitor of DYRK, with IC50s of 0.36 nM (DYRK1A) and 0.59 nM (DYRK1B). It exhibits antileukemia effect, regulates cell cycle, and induces cell apoptosis.
  • Induces apoptosis of primary ALL cells that were resistant to cytarabine.
  • Dose-dependently induces apoptosis in B- and T-cell lines and primary human pediatric cells.
  • Inhibits DYRK1A, leading to a loss of DYRK1A-mediated FOXO1 and STAT3 signaling, resulting in preferential cell death in leukemic B cells.
  • Inhibits phosphorylation of FOXO1, STAT3, and cyclin D3, regulating late cell-cycle progression, mitochondrial ROS, and DNA damage.
  • Demonstrates antileukemia activity in a leukemic aggressive model in mice, reducing leukemic burden and conferring a modest survival advantage.
Catalog No. 50-003-43327 Supplier Medchemexpress LLC Supplier No. HY11138050MG
EHT 1610 is a potent inhibitor of DYRK, with IC50s of 0.36 nM (DYRK1A) and 0.59 nM (DYRK1B). It exhibits antileukemia effect, regulates cell cycle, and induces cell apoptosis.
  • Induces apoptosis of primary ALL cells that were resistant to cytarabine.
  • Dose-dependently induces apoptosis in B- and T-cell lines and primary human pediatric cells.
  • Inhibits DYRK1A, leading to a loss of DYRK1A-mediated FOXO1 and STAT3 signaling, resulting in preferential cell death in leukemic B cells.
  • Inhibits phosphorylation of FOXO1, STAT3, and cyclin D3, regulating late cell-cycle progression, mitochondrial ROS, and DNA damage.
  • Demonstrates antileukemia activity in a leukemic aggressive model in mice, reducing leukemic burden and conferring a modest survival advantage.