Learn More
Selleck Chemical LLC Thiazolyl Blue
SDP
Supplier:  Selleck Chemical LLC S6821100MG
Encompass_Preferred
Thiazolyl Blue (MTT Tetrazolium bromide Methylthiazolyldiphenyl-tetrazolium bromide Methylthialazole Tetrazolium) is a cell-permeable and positively charged colorimetric agent that is widely used to detect reductive metabolism in cells and to measure cell proliferation cytotoxicity and apoptosis Thiazolyl Blue (MTT) is reduced from yellow color to purple formazan in living cells
Catalog No. 50-000-02526