Learn More
AdipoGen Thiazolyl blue tetrazolium Br
SDP
Catalog No. 502559970
Encompass_Preferred
Chemical. CAS 298-93-1. Formula C18H16BrN5S. MW 414.32. MTT is a common histochemical/cytochemical reagent and for the detection of NAD. MTT is used to detect reductive metabolism in cells. It is a cell-permeable yellow dye that is reduced by intracellular NADPH-oxidoreductases in mitochondria of living cells to form the dark blue water-insoluble MTT-formazan. The intensity of the color can then be measured spectrophotometrically at 540 or 570nm, which allows quantitation of cell viability and cell proliferation. It is frequently used in colorimetric assays to measure cell proliferation/cell growth, cytotoxicity such as for screening cancer drugs, and apoptosis.
Catalog No. 50-255-9970 Supplier AdipoGen Supplier No. CDXT0186G001
Chemical. CAS 298-93-1. Formula C18H16BrN5S. MW 414.32. MTT is a common histochemical/cytochemical reagent and for the detection of NAD. MTT is used to detect reductive metabolism in cells. It is a cell-permeable yellow dye that is reduced by intracellular NADPH-oxidoreductases in mitochondria of living cells to form the dark blue water-insoluble MTT-formazan. The intensity of the color can then be measured spectrophotometrically at 540 or 570nm, which allows quantitation of cell viability and cell proliferation. It is frequently used in colorimetric assays to measure cell proliferation/cell growth, cytotoxicity such as for screening cancer drugs, and apoptosis.