Learn More
Abcam Thiazovivin (DMSO solution), Rho kinase inhibitor, 1MG
Supplier:  Abcam AB1466181MG
Encompass_Preferred
MW 311.4 Da, Purity >95%. Rho kinase inhibitor. Thiazovivin (ab142081) solution. Reverses Ang II-induced MYPT1 and MLC phosphorylation. Enhances fibroblast reprogramming efficiency during pluripotent stem cell formation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5325