Learn More
Abcam Thiazovivin, ROCK inhibitor, 1MG
Catalog No. 502706345
Encompass_Preferred
MW 311.4 Da, Purity >98%. Rho kinase inhibitor. Thiazovivin (ab142081) solution. Reverses Ang II-induced MYPT1 and MLC phosphorylation. Enhances fibroblast reprogramming efficiency during pluripotent stem cell formation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6345 Supplier Abcam Supplier No. AB1420811MG
MW 311.4 Da, Purity >98%. Rho kinase inhibitor. Thiazovivin (ab142081) solution. Reverses Ang II-induced MYPT1 and MLC phosphorylation. Enhances fibroblast reprogramming efficiency during pluripotent stem cell formation.
The product is subject to the following: Abcam Restricted Use Statement