Learn More
MSC AbilityOne Floor Pad, Nylon, Buffing, Tan, 19 in Diameter
Catalog No. 502839537
Encompass
These nylon floor pads, with a 19 in diameter, are designed for use with concentrated weight, rotary disk floor machines operating between 150-350 rpm. They are washable, reusable, and corrosion-resistant. Suitable for all resilient tile, terrazzo, marble, and ceramic floors, these pads effectively remove light scuffs and dirt, revitalizing floor appearance.
  • Effectively removes light scuffs and dirt
  • Revitalizes the appearance of floors
  • Safe for most types of floors
  • Cleans when wet, buffs when dry
  • Conforms to the floor's contours, ensuring thorough cleaning and buffing
Catalog No. 50-283-9537 Supplier MSC Supplier No. 89461172
These nylon floor pads, with a 19 in diameter, are designed for use with concentrated weight, rotary disk floor machines operating between 150-350 rpm. They are washable, reusable, and corrosion-resistant. Suitable for all resilient tile, terrazzo, marble, and ceramic floors, these pads effectively remove light scuffs and dirt, revitalizing floor appearance.
  • Effectively removes light scuffs and dirt
  • Revitalizes the appearance of floors
  • Safe for most types of floors
  • Cleans when wet, buffs when dry
  • Conforms to the floor's contours, ensuring thorough cleaning and buffing