Learn More
MSC AbilityOne Floor Pad, Nylon, Stripping, Black, 20 in Diameter
Catalog No. 502836160
Encompass
These floor pads are for use on concentrated weight, rotary disk floor machines rated between 150 - 350 rpm. They are washable, reusable, and corrosion-resistant. The pads have a 3-3/8 in center hole and are 11/16 in thick. They are suitable for use on all resilient tile, terrazzo, marble, and ceramic floors. Each box contains 5 pads.
  • Used with water-based floor finish stripping solutions
  • Cleans heavily soiled surface floors such as concrete
  • Made of nylon and polyester fibers
  • Washable
  • Use with low speed 175 - 600 RPM floor machines
Catalog No. 50-283-6160 Supplier MSC Supplier No. 89461206
These floor pads are for use on concentrated weight, rotary disk floor machines rated between 150 - 350 rpm. They are washable, reusable, and corrosion-resistant. The pads have a 3-3/8 in center hole and are 11/16 in thick. They are suitable for use on all resilient tile, terrazzo, marble, and ceramic floors. Each box contains 5 pads.
  • Used with water-based floor finish stripping solutions
  • Cleans heavily soiled surface floors such as concrete
  • Made of nylon and polyester fibers
  • Washable
  • Use with low speed 175 - 600 RPM floor machines