Learn More
Sutter Instruments THICK/STANDARD WALL BOROSILICA
SDP
Catalog No. NC1930508
Encompass_Preferred
Thick/Standard Wall Borosilicate Glass Outside Diameter: 1.00 mm Inside Diameter: 0.50 mm Overall Length :10 cm Pieces per Package 225ea
Catalog No. NC1930508 Supplier Sutter Instruments Supplier No. FGGBF1005010
Thick/Standard Wall Borosilicate Glass Outside Diameter: 1.00 mm Inside Diameter: 0.50 mm Overall Length :10 cm Pieces per Package 225ea