Learn More
McMaster-Carr Thick-Wall 316/316L Stainless Steel Pipe Nipple
Catalog No. NC1788123
Encompass
Threaded on Both Ends, 3/4 Pipe Size, 6" Long. Thick walls, also known as Schedule 80, give this pipe the strength to handle high-pressure applications. It is seamless so it has a smooth interior for unrestricted flow. 316 stainless steel pipe has excellent corrosion resistance.
Catalog No. NC1788123 Supplier McMaster-Carr Supplier No. 4475K75
Threaded on Both Ends, 3/4 Pipe Size, 6" Long. Thick walls, also known as Schedule 80, give this pipe the strength to handle high-pressure applications. It is seamless so it has a smooth interior for unrestricted flow. 316 stainless steel pipe has excellent corrosion resistance.