Learn More
Cayman Chemical Thidiazuron, Purity: ≥98%, CAS: 51707-55-2, 50 mg
SDP
Catalog No. 502980616
Encompass_Preferred
A plant growth regulator; increases soybean callus and radish cotyledon growth at 5 nM and 50 µM, respectively; induces the dedifferentiation of, and increases seedling number from, cultured geranium hypocotyls at 400 nM for seven weeks; induces defoliation in cotton in greenhouse studies at 56 g/ha; reduces the viability of MDA-MB-231 and MCF-7 cell lines in a concentration-dependent manner, as well as increases DNA fragmentation and apoptosis in MDA-MB-231 cells at 50 µM; increases E-cadherin and decreases vimentin levels in, as well as reduces the number of mammospheres formed by, MDA-MB-231 cells at 50 µM
Catalog No. 50-298-0616 Supplier Cayman Chemical Supplier No. 1727250 MG
A plant growth regulator; increases soybean callus and radish cotyledon growth at 5 nM and 50 µM, respectively; induces the dedifferentiation of, and increases seedling number from, cultured geranium hypocotyls at 400 nM for seven weeks; induces defoliation in cotton in greenhouse studies at 56 g/ha; reduces the viability of MDA-MB-231 and MCF-7 cell lines in a concentration-dependent manner, as well as increases DNA fragmentation and apoptosis in MDA-MB-231 cells at 50 µM; increases E-cadherin and decreases vimentin levels in, as well as reduces the number of mammospheres formed by, MDA-MB-231 cells at 50 µM