Learn More
Medchemexpress LLC Thidiazuron | 51707-55-2 | MFCD00078723 | 99.7% | 1 ML
SDP
Catalog No. 5000407690
Encompass_Preferred
Thidiazuron is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It functions as a plant growth regulator, often utilized as a supplement in media like Murashige and Skoog for micropropagation. This reagent is known for promoting plant organogenesis, including bud regeneration, and overall plant regeneration.
  • Used as a supplement to media for micropropagation.
  • Promotes plant organogenesis (bud regeneration).
  • Promotes plant regeneration.
  • High purity.
Catalog No. 50-004-07690 Supplier Medchemexpress LLC Supplier No. HYB087210MM/1ML
Thidiazuron is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It functions as a plant growth regulator, often utilized as a supplement in media like Murashige and Skoog for micropropagation. This reagent is known for promoting plant organogenesis, including bud regeneration, and overall plant regeneration.
  • Used as a supplement to media for micropropagation.
  • Promotes plant organogenesis (bud regeneration).
  • Promotes plant regeneration.
  • High purity.