Learn More
Cayman Chemical ThIelavIn A 2.5mg
SDP
Catalog No. 502987379
Encompass_Preferred
A natural COX inhibitor; blocks conversion of arachidonic acid to PGH2 (IC50 = 10 µM); blocks conversion of PGH2 to PGE2 (IC50 = 40 µM); inhibits glucose-6-phosphatase (IC50 = 4.6 µM); non-competitively inhibits α-glucosidase (IC50 = 23.8 µM; Ki = 27.8 µM)
Catalog No. 50-298-7379 Supplier Cayman Chemical Supplier No. 217672.5 MG
Add to Cart
Add to Cart
A natural COX inhibitor; blocks conversion of arachidonic acid to PGH2 (IC50 = 10 µM); blocks conversion of PGH2 to PGE2 (IC50 = 40 µM); inhibits glucose-6-phosphatase (IC50 = 4.6 µM); non-competitively inhibits α-glucosidase (IC50 = 23.8 µM; Ki = 27.8 µM)