Learn More
Cayman Chemical ThIelavIn B 2.5mg
SDP
Catalog No. 502980154
Encompass_Preferred
A fungal metabolite that inhibits cyclooxygenase, blocking both the conversion of AA to PGH2 and the conversion of PGH2 to PGE2 (IC50s = 40 and 9 µM, respectively); also inhibits the reverse transcriptase of avian myeloblastosis virus, bacterial transglycosylases, and telomerase activity
Catalog No. 50-298-0154 Supplier Cayman Chemical Supplier No. 187702.5 MG
A fungal metabolite that inhibits cyclooxygenase, blocking both the conversion of AA to PGH2 and the conversion of PGH2 to PGE2 (IC50s = 40 and 9 µM, respectively); also inhibits the reverse transcriptase of avian myeloblastosis virus, bacterial transglycosylases, and telomerase activity