Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thieno[2,3-b]pyrazin-7-amine, Thermo Scientific™™

Catalog No. MO07822DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07822DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07822DA Supplier Thermo Scientific Chemicals Supplier No. MO07822DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59944-75-1
Molecular Formula C6H5N3S
Molecular Weight (g/mol) 151.187
MDL Number MFCD08271970
InChI Key ZRVKSPNBHZCQKY-UHFFFAOYSA-N
Synonym thieno 2,3-b pyrazin-7-amine, thieno 3,2-b pyrazin-7-amine, 7-aminothieno 2,3-b pyrazine, 4ctk, svn, 7-aminothieno 2,3-b pyrazin, thiopheno 3,2-b pyrazine-7-ylamine
PubChem CID 12299130
IUPAC Name thieno[2,3-b]pyrazin-7-amine
SMILES C1=CN=C2C(=N1)C(=CS2)N

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 151.1858
Chemical Name or Material Thieno[2, 3-b]pyrazin-7-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.