Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thieno[2,3-c]isoquinolin-5(4H)-one | 420849-22-5 | 99.9% | 1 ML
SDP

Catalog No. 5000232467
Encompass_Preferred

TIQ-A (thieno[2,3-c]isoquinolin-5(4H)-one) is a small-molecule tankyrase (TNKS2) inhibitor used in research; it potently inhibits TNKS2 (IC50 = 24 nM) and shows activity against multiple PARP-family enzymes.

  • Potent TNKS2 inhibition (IC50 24 nM) and activity against PARP2 and PARP15.
  • High listed purity: 99.85%.
  • Molecular formula C11H7NOS; molecular weight 201.24.
  • Available as 10 mM solution in DMSO (1 ml) and as powder in sizes from 1 mg to 100 mg.
  • DMSO solubility reported at 125 mg/mL; ultrasonic agitation may be required.
  • Storage: powder at -20°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-32467 Supplier Medchemexpress LLC Supplier No. HY12171910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TIQ-A (thieno[2,3-c]isoquinolin-5(4H)-one) is a small-molecule tankyrase (TNKS2) inhibitor used in research; it potently inhibits TNKS2 (IC50 = 24 nM) and shows activity against multiple PARP-family enzymes.

  • Potent TNKS2 inhibition (IC50 24 nM) and activity against PARP2 and PARP15.
  • High listed purity: 99.85%.
  • Molecular formula C11H7NOS; molecular weight 201.24.
  • Available as 10 mM solution in DMSO (1 ml) and as powder in sizes from 1 mg to 100 mg.
  • DMSO solubility reported at 125 mg/mL; ultrasonic agitation may be required.
  • Storage: powder at -20°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.