Learn More
Medchemexpress LLC Thieno[2,3-c]pyridine-6(5H)-carboxamide, 3-cyano-4,7-dihydro-N-(2-methoxyphenyl)-2-[[(1-methyl-1H-pyrazol-4-yl)carbonyl]amino]- | 2248552-84-1 | 99.9% | 436.49 | 1 ML
SDP
Catalog No. 5000392368
Encompass_Preferred
WB436B is a highly selective STAT3 inhibitor that can selectively inhibit STAT3-Tyr705 phosphorylation and the expression of STAT3 target genes. It shows cytotoxic effects on pancreatic cancer cells, induces apoptosis, and can suppress tumor growth and metastasis in pancreatic cancer mouse models, extending the survival of tumor-bearing mice.
  • Selectively inhibits STAT3-Tyr705 phosphorylation
  • Suppresses expression of STAT3 target genes
  • Induces apoptosis in pancreatic cancer cells
  • Suppresses tumor growth and metastasis in pancreatic cancer mouse models
  • Extends survival in tumor-bearing mice
Catalog No. 50-003-92368 Supplier Medchemexpress LLC Supplier No. HY16454610MM/1ML
WB436B is a highly selective STAT3 inhibitor that can selectively inhibit STAT3-Tyr705 phosphorylation and the expression of STAT3 target genes. It shows cytotoxic effects on pancreatic cancer cells, induces apoptosis, and can suppress tumor growth and metastasis in pancreatic cancer mouse models, extending the survival of tumor-bearing mice.
  • Selectively inhibits STAT3-Tyr705 phosphorylation
  • Suppresses expression of STAT3 target genes
  • Induces apoptosis in pancreatic cancer cells
  • Suppresses tumor growth and metastasis in pancreatic cancer mouse models
  • Extends survival in tumor-bearing mice