Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thieno[2,3-c]pyridine, 7-chloro- | 28948-58-5 | 99.4% | 169.63 | C7H4ClNS | 500 MG
SDP

Catalog No. 5000209949
Encompass_Preferred

7-Chlorothieno[2,3-c]pyridine is an aromatic heterocyclic compound used as a synthetic intermediate in pharmaceutical and medicinal chemistry research. It is typically supplied as a high-purity solid for use in organic synthesis and R&D.

  • Used as a synthetic intermediate in pharmaceutical synthesis.
  • High purity (99.4% by HPLC).
  • Molecular weight 169.63.
  • CAS number 28948-58-5.
  • Available in small-scale packages suitable for research (250 mg-5 g; larger quantities by inquiry).

Catalog No. 50-002-09949 Supplier Medchemexpress LLC Supplier No. HYW058182500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Chlorothieno[2,3-c]pyridine is an aromatic heterocyclic compound used as a synthetic intermediate in pharmaceutical and medicinal chemistry research. It is typically supplied as a high-purity solid for use in organic synthesis and R&D.

  • Used as a synthetic intermediate in pharmaceutical synthesis.
  • High purity (99.4% by HPLC).
  • Molecular weight 169.63.
  • CAS number 28948-58-5.
  • Available in small-scale packages suitable for research (250 mg-5 g; larger quantities by inquiry).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.