Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thieno[2,3-c]pyridine, 7-chloro | 28948-58-5 | MFCD04973300 | 99.4% | 169.63 g/mol | C7H4ClNS | 250 MG
SDP

Catalog No. 5000209662
Encompass_Preferred

7-Chlorothieno[2,3-c]pyridine is a heterocyclic synthetic intermediate used in pharmaceutical and medicinal chemistry synthesis. It is a white to light yellow solid with a molecular weight of 169.63 g/mol and a molecular formula of C7H4ClNS. The compound is commonly supplied in small quantities for research and development and is used as a building block for heterocycle construction.

  • High purity (99.4% by HPLC).
  • Molecular formula C7H4ClNS.
  • Molecular weight 169.63 g/mol.
  • White to light yellow solid appearance.
  • CAS number 28948-58-5.
  • Commonly supplied in 250 mg quantities for research use.

Catalog No. 50-002-09662 Supplier Medchemexpress LLC Supplier No. 250MGHYW058182
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

7-Chlorothieno[2,3-c]pyridine is a heterocyclic synthetic intermediate used in pharmaceutical and medicinal chemistry synthesis. It is a white to light yellow solid with a molecular weight of 169.63 g/mol and a molecular formula of C7H4ClNS. The compound is commonly supplied in small quantities for research and development and is used as a building block for heterocycle construction.

  • High purity (99.4% by HPLC).
  • Molecular formula C7H4ClNS.
  • Molecular weight 169.63 g/mol.
  • White to light yellow solid appearance.
  • CAS number 28948-58-5.
  • Commonly supplied in 250 mg quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.