Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thieno[3,2-b]thiophene-2-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. T31561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31561G 1 g
T3156200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31561G Supplier TCI America Supplier No. T31561G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1723-27-9
Molecular Formula C7H4O2S2
Molecular Weight (g/mol) 184.23
MDL Number MFCD00115174
InChI Key GVZXSZWCZGKLRS-UHFFFAOYSA-N
Synonym thieno 3,2-b thiophene-2-carboxylic acid, thieno 3,2-b thiophene-5-carboxylic acid, thieno 3,2-b thiophene-2-carboxylicacid, thiopheno 3,2-b thiophene-2-carboxylic acid, zlchem 802, maybridge3_006605, thienothiophene carboxylic acid, 5-thieno 3,2-b thiophenecarboxylic acid, nuk
PubChem CID 660319
IUPAC Name thieno[3,2-b]thiophene-2-carboxylic acid
SMILES OC(=O)C1=CC2=C(S1)C=CS2

Specifications

Melting Point 223°C
Color Yellow
Quantity 1 g
Formula Weight 184.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Thieno[3,2-b]thiophene-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.