Learn More
Medchemexpress LLC Thieno[3,2-c]pyridine-5(4H)-acetic acid, α-(2-chlorophenyl)-6,7-dihydro-, hydrochloride | 144750-42-5 | MFCD08063656 | 99.7% | 344.26 | C15H15Cl2NO2S | 5 MG
SDP
Catalog No. 5000221484
Encompass_Preferred
Clopidogrel carboxylic acid hydrochloride is the hydrochloride salt of the inactive primary metabolite of clopidogrel, provided as a research-use reference standard for analytical, pharmacokinetic, and metabolic studies (CAS 144750-42-5). The material is supplied in small milligram quantities with batch documentation to support analytical traceability.
  • Provided as a research-use reference standard for analytical assays.
  • High stated purity for accurate quantitation and method development.
  • Supplied in small milligram pack sizes suitable for laboratory use.
  • Hydrochloride salt improves stability and handling in many assays.
  • Accompanied by batch-specific certificate of analysis.
Catalog No. 50-002-21484 Supplier Medchemexpress LLC Supplier No. HY141749B5MG
Clopidogrel carboxylic acid hydrochloride is the hydrochloride salt of the inactive primary metabolite of clopidogrel, provided as a research-use reference standard for analytical, pharmacokinetic, and metabolic studies (CAS 144750-42-5). The material is supplied in small milligram quantities with batch documentation to support analytical traceability.
  • Provided as a research-use reference standard for analytical assays.
  • High stated purity for accurate quantitation and method development.
  • Supplied in small milligram pack sizes suitable for laboratory use.
  • Hydrochloride salt improves stability and handling in many assays.
  • Accompanied by batch-specific certificate of analysis.