Learn More
Apexbio Technology LLC Thieno-CTP 50ul (100 mM)
SDP
Supplier:  Apexbio Technology LLC B80650.05
Encompass_Preferred
Thieno-CTP is a structural analog of cytidine triphosphate (CTP) that incorporates a thiophene-modified nucleobase allowing incorporation into RNA by RNA polymerases during transcription Following enzymatic incorporation into nascent RNA transcripts the introduced thienyl modification acts as a reactive site suitable for subsequent bioorthogonal labeling via selective chemical reactions This chemical feature enables detection visualization and analysis of newly synthesized RNA molecules within biological systems Hence Thieno-CTP is utilized to examine transcriptional dynamics RNA synthetic regulation RNA trafficking and RNA-protein interaction studies supporting research into RNA biology and related areas
Catalog No. 50-000-63804