Learn More
Medchemexpress LLC Thiethylperazine dimaleate | 1179-69-7 | 10 MG
SDP
Catalog No. 5000295821
Encompass_Preferred
Thiethylperazine dimaleate is a phenothiazine derivative used as a research reagent and analytical standard. It acts as a dopamine D2 and histamine H1 receptor antagonist, and has demonstrated anti-emetic, antipsychotic, and antimicrobial activities; it has also been reported to activate ABCC1 and reduce amyloid-β load in animal studies.
  • Dopamine D2 and histamine H1 receptor antagonist.
  • Anti-emetic and antipsychotic activity reported in studies.
  • ABCC1 activator shown to reduce amyloid-β in mice.
  • Molecular weight 631.76 g/mol; CAS 1179-69-7.
  • Supplied as the dimaleate salt; store sealed at 4°C, protect from moisture and light.
Catalog No. 50-002-95821 Supplier Medchemexpress LLC Supplier No. HYB1794A10MG
Thiethylperazine dimaleate is a phenothiazine derivative used as a research reagent and analytical standard. It acts as a dopamine D2 and histamine H1 receptor antagonist, and has demonstrated anti-emetic, antipsychotic, and antimicrobial activities; it has also been reported to activate ABCC1 and reduce amyloid-β load in animal studies.
  • Dopamine D2 and histamine H1 receptor antagonist.
  • Anti-emetic and antipsychotic activity reported in studies.
  • ABCC1 activator shown to reduce amyloid-β in mice.
  • Molecular weight 631.76 g/mol; CAS 1179-69-7.
  • Supplied as the dimaleate salt; store sealed at 4°C, protect from moisture and light.