Learn More
Medchemexpress LLC Thiethylperazine (dimaleate) | 1179-69-7 | 99.99% | 100 MG
SDP
Catalog No. 5000412277
Encompass_Preferred
Thiethylperazine dimaleate is a phenothiazine derivative, functioning as an orally active and potent dopamine D2-receptor and histamine H1-receptor antagonist. It is also a selective ABCC1 activator that reduces amyloid-β (Aβ) load in mice. It possesses anti-emetic, antipsychotic, and antimicrobial effects.
  • Potent dopamine D2-receptor antagonist
  • Histamine H1-receptor antagonist
  • Selective ABCC1 activator
  • Reduces amyloid-β (Aβ) load in mice
  • Anti-emetic, antipsychotic, and antimicrobial effects
  • Enhances antibiotic (Vancomycin) activity
Catalog No. 50-004-12277 Supplier Medchemexpress LLC Supplier No. HYB1794A100MG
Thiethylperazine dimaleate is a phenothiazine derivative, functioning as an orally active and potent dopamine D2-receptor and histamine H1-receptor antagonist. It is also a selective ABCC1 activator that reduces amyloid-β (Aβ) load in mice. It possesses anti-emetic, antipsychotic, and antimicrobial effects.
  • Potent dopamine D2-receptor antagonist
  • Histamine H1-receptor antagonist
  • Selective ABCC1 activator
  • Reduces amyloid-β (Aβ) load in mice
  • Anti-emetic, antipsychotic, and antimicrobial effects
  • Enhances antibiotic (Vancomycin) activity