Learn More
Medchemexpress LLC Thiethylperazine dimaleate | 1179-69-7 | 99.99% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1794A50MG
Encompass_Preferred
Thiethylperazine dimaleate is a phenothiazine derivative that is an orally active and potent dopamine D2-receptor and histamine H1-receptor antagonist. It is also a selective ABCC1 activator that reduces amyloid-β (Aβ) load in mice. It possesses anti-emetic, antipsychotic, and antimicrobial effects.
  • Orally active and potent dopamine D2-receptor and histamine H1-receptor antagonist.
  • Selective ABCC1 activator.
  • Reduces amyloid-β (Aβ) load in mice.
  • Anti-emetic, antipsychotic, and antimicrobial effects.
Catalog No. 50-004-12779