Learn More
AdipoGen Thifluzamide
SDP
Catalog No. 502559997
Encompass_Preferred
Chemical. CAS 130000-40-7. Formula C13H6Br2F6N2O2S. MW 528.1. Synthetic. Thiazole fungicide used in agrochemistry. Succinate dehydrogenase inhibitor SDHI which interferes with succinate ubiquinone reductase in the mitochondrial electron transport chain of fungi.Compound can be used as analytical reference material.
Catalog No. 50-255-9997 Supplier AdipoGen Supplier No. CDXT0118M050
Chemical. CAS 130000-40-7. Formula C13H6Br2F6N2O2S. MW 528.1. Synthetic. Thiazole fungicide used in agrochemistry. Succinate dehydrogenase inhibitor SDHI which interferes with succinate ubiquinone reductase in the mitochondrial electron transport chain of fungi.Compound can be used as analytical reference material.